Octahydrocyclopenta[c]pyrrole 98%
Supplier: Apollo Scientific
OR916903-10G
OR916903-1G
OR916903-25G
APOSOR916903-10GEA
348
EUR
APOSOR916903-10G
APOSOR916903-1G
APOSOR916903-25G
Octahydrocyclopenta[c]pyrrole 98%
Octahydrocyclopenta[c]pyrrole
Formula:
C₇H₁₃N Storage Temperature: Ambient |
CAS Number:
5661-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...