4-Chloro-2-isopropyl-5-methylphenol 98%

Supplier: Apollo Scientific

Synonyms: 6-Chlorothymol

OR916910-100G OR916910-25G
APOSOR916910-100GEA 328 EUR
APOSOR916910-100G APOSOR916910-25G
4-Chloro-2-isopropyl-5-methylphenol 98%
4-Chloro-2-isopropyl-5-methylphenol
Formula: C₁₀H₁₃ClO
MW: 184,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002326
CAS Number: 89-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR