4-Methyl-1-phenyl-2-pentanol 96%

Supplier: Apollo Scientific

OR917429-25G OR917429-5G
APOSOR917429-25GEA 127 EUR
APOSOR917429-25G APOSOR917429-5G
4-Methyl-1-phenyl-2-pentanol 96%
4-Methyl-1-phenyl-2-pentanol
Formula: C₁₂H₁₈O
MW: 178,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209515
CAS Number: 7779-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR