Tetraethylurea 95%
Supplier: Apollo Scientific
OR917459-100G
OR917459-25G
APOSOR917459-100GEA
435
EUR
APOSOR917459-100G
APOSOR917459-25G
Tetraethylurea 95%
Tetraethylurea
Formula:
C₉H₂₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD00042881 CAS Number: 1187-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...