N-Acetyl-DL-norleucine 95%
Supplier: Apollo Scientific
Synonyms:
Ac-DL-Nle-OH
OR917582-25G
OR917582-5G
OR917582-100G
APOSOR917582-25GEA
256
EUR
APOSOR917582-25G
APOSOR917582-5G
APOSOR917582-100G
N-Acetyl-DL-norleucine 95%
N-Acetyl-DL-norleucine
Formula:
C₈H₁₅NO₃ MW: 173,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037272 CAS Number: 7682-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...