2-Hydroxybenzophenone 95%

Supplier: Apollo Scientific

Synonyms: (2-Hydroxyphenyl)(phenyl)methanone, o-Hydroxybenzophenone

OR917799-25G OR917799-5G
APOSOR917799-25GEA 221 EUR
APOSOR917799-25G APOSOR917799-5G
2-Hydroxybenzophenone 95%
2-Hydroxybenzophenone
Formula: C₁₃H₁₀O₂
MW: 198,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002216
CAS Number: 117-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR