2-Oxo-2H-chromene-6-carboxylic acid 95%

Supplier: Apollo Scientific

OR918069-1G OR918069-250MG OR918069-5G
APOSOR918069-1GEA 248 EUR
APOSOR918069-1G APOSOR918069-250MG APOSOR918069-5G
2-Oxo-2H-chromene-6-carboxylic acid 95%
2-Oxo-2H-chromene-6-carboxylic acid
Formula: C₁₀H₆O₄
Storage Temperature: Ambient
MDL Number: MFCD06203567
CAS Number: 7734-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR