2-Bromo-6-pyrrolidin-1-ylpyridine

Supplier: Apollo Scientific

Synonyms: 2-Bromo-6-(pyrrolidin-1-yl)pyridine

OR9181-1G OR9181-5G
APOSOR9181-1GEA 55.3 EUR
APOSOR9181-1G APOSOR9181-5G
2-Bromo-6-pyrrolidin-1-ylpyridine
2-Bromo-6-pyrrolidin-1-ylpyridine
Formula: C₉H₁₁BrN₂
MW: 227,11 g/mol
Melting Pt: 86…88 °C
Storage Temperature: Ambient
MDL Number: MFCD08271892
CAS Number: 230618-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR