2-Methoxybenzenesulphonamide 95%

Supplier: Apollo Scientific

OR918215-1G OR918215-2G
APOSOR918215-1GEA 524 EUR
APOSOR918215-1G APOSOR918215-2G
2-Methoxybenzenesulphonamide 95%
2-Methoxybenzenesulphonamide
Formula: C₇H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD06496492
CAS Number: 52960-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR