N,N-Dimethyl-His-OMe 95%
Supplier: Apollo Scientific
OR918382-1G
OR918382-250MG
APOSOR918382-1GEA
864
EUR
APOSOR918382-1G
APOSOR918382-250MG
N,N-Dimethyl-His-OMe 95%
N,N-Dimethyl-His-OMe
Formula:
C₉H₁₅N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01318773 CAS Number: 170227-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...