N-Methyl-N-[(3-methyl-1-propyl-1H-pyrazol-5-yl)methyl]amine 95%

OR918542-1G OR918542-250MG
APOSOR918542-1GEA 684 EUR
APOSOR918542-1G APOSOR918542-250MG
N-Methyl-N-[(3-methyl-1-propyl-1H-pyrazol-5-yl)methyl]amine 95%
N-Methyl-N-[(3-methyl-1-propyl-1H-pyrazol-5-yl)methyl]amine
Formula: C₉H₁₇N₃
MW: 167,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08277101
CAS Number: 956208-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR