4-Bromo-2-methylpyridine 98%
Supplier: Apollo Scientific
|
Warning
|
OR9191-100G
OR9191-25G
OR9191-5G
OR9191-1G
APOSOR9191-100GEA
190
EUR
APOSOR9191-100G
APOSOR9191-25G
APOSOR9191-5G
APOSOR9191-1G
4-Bromo-2-methylpyridine 98%
4-Bromo-2-methylpyridine
Formula:
C₆H₆BrN MW: 172,02 g/mol Density: 1,45 g/cm³ (25 °C) Flash Pt: 78,9 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788196 CAS Number: 22282-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...