2,6-Dimethyl-p-benzoquinone 95%

Supplier: Apollo Scientific

Synonyms: 2,6-Dimethylcyclohexa-2,5-diene-1,4-dione

OR919100-5G
APOSOR919100-5GEA 63.8 EUR
APOSOR919100-5G
2,6-Dimethyl-p-benzoquinone 95%
2,6-Dimethyl-p-benzoquinone
Formula: C₈H₈O₂
MW: 136,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001605
CAS Number: 527-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR