Ethyl-2-formyl-3-oxopropionate 96%

Supplier: Apollo Scientific
Warning

OR919533-100G OR919533-25G
APOSOR919533-100GEA 201 EUR
APOSOR919533-100G APOSOR919533-25G
Ethyl-2-formyl-3-oxopropionate 96%
Ethyl-2-formyl-3-oxopropionate
Formula: C₆H₈O₄
Storage Temperature: Refrigerator
MDL Number: MFCD11112084
CAS Number: 80370-42-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR