3-Methyl-2-oxazolidone 97%

Supplier: Apollo Scientific

Synonyms: 2-Oxo-3-methyloxazolidine

OR919648-100G OR919648-25G OR919648-500G
APOSOR919648-100GEA 139 EUR
APOSOR919648-100G APOSOR919648-25G APOSOR919648-500G
3-Methyl-2-oxazolidone 97%
3-Methyl-2-oxazolidone
Formula: C₄H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00009759
CAS Number: 19836-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR