Isoamyl lactate 95%
Supplier: Apollo Scientific
OR919725-25G
OR919725-5G
APOSOR919725-25GEA
284
EUR
APOSOR919725-25G
APOSOR919725-5G
Isoamyl lactate 95%
Isoamyl lactate
Formula:
C₈H₁₆O₃ MW: 160,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046638 CAS Number: 19329-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...