1,2,4,5-Tetramethylimidazole 95%

Supplier: Apollo Scientific

Synonyms: 1,2,4,5-Tetramethyl-1H-imidazole

OR919739-1G OR919739-25G OR919739-5G
APOSOR919739-1GEA 21.9 EUR
APOSOR919739-1G APOSOR919739-25G APOSOR919739-5G
1,2,4,5-Tetramethylimidazole 95%
1,2,4,5-Tetramethylimidazole
Formula: C₇H₁₂N₂
MW: 124,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059162
CAS Number: 1739-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR