2,3-Dichlorobenzyl alcohol 95%

Supplier: Apollo Scientific

Synonyms: (2,3-Dichlorophenyl)methanol

OR919849-10G OR919849-1G OR919849-5G
APOSOR919849-10GEA 553 EUR
APOSOR919849-10G APOSOR919849-1G APOSOR919849-5G
2,3-Dichlorobenzyl alcohol 95%
2,3-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238581
CAS Number: 38594-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR