1-Bromo-4-chlorobutane 97+%
Supplier: Apollo Scientific
|
Warning
|
OR919950-100G
OR919950-500G
APOSOR919950-100GEA
147
EUR
APOSOR919950-100G
APOSOR919950-500G
1-Bromo-4-chlorobutane 97+%
1-Bromo-4-chlorobutane
Formula:
BrH₂C(CH₂)₂CH₂Cl MW: 171.46 g/mol Boiling Pt: 175…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001010 CAS Number: 6940-78-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...