(±)-1-Heptyn-3-ol 97%

Supplier: Apollo Scientific

Synonyms: Hept-1-yn-3-ol

OR920099-1G OR920099-5G
APOSOR920099-1GEA 118 EUR
APOSOR920099-1G APOSOR920099-5G
(±)-1-Heptyn-3-ol 97%
(±)-1-Heptyn-3-ol
Formula: C₇H₁₂O
MW: 112,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041597
CAS Number: 7383-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR