2-(Methylthio)benzothiazole 97%

Supplier: Apollo Scientific

Synonyms: 2-(Methylthio)benzo[d]thiazole

OR920222-100G OR920222-500G
APOSOR920222-100GEA 82.8 EUR
APOSOR920222-100G APOSOR920222-500G
2-(Methylthio)benzothiazole 97%
2-(Methylthio)benzothiazole
Formula: C₈H₇NS₂
MW: 181,2779 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005784
CAS Number: 615-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR