Allyl isonicotinate 95%
Supplier: Apollo Scientific
Synonyms:
Allyl 4-Pyridinecarboxylate, 4-Pyridinecarboxylic Acid Allyl Ester, Isonicotinic Acid Allyl Ester
OR920386-1G
OR920386-25G
OR920386-5G
APOSOR920386-1GEA
161
EUR
APOSOR920386-1G
APOSOR920386-25G
APOSOR920386-5G
Allyl isonicotinate 95%
Allyl isonicotinate
Formula:
C₉H₉NO₂ MW: 163,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059786 CAS Number: 25635-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...