4-Methoxy-3-(methoxymethyl)benzaldehyde 95%

Supplier: Apollo Scientific

OR920553-1G OR920553-5G
APOSOR920553-1GEA 78.3 EUR
APOSOR920553-1G APOSOR920553-5G
4-Methoxy-3-(methoxymethyl)benzaldehyde 95%
4-Methoxy-3-(methoxymethyl)benzaldehyde
Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01822195
CAS Number: 76646-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR