N,N-Diethylacetamide 95%
Supplier: Apollo Scientific
Synonyms:
N-Acetyldiethylamine
OR920831-25G
OR920831-500G
APOSOR920831-25GEA
52.4
EUR
APOSOR920831-25G
APOSOR920831-500G
N,N-Diethylacetamide 95%
N,N-Diethylacetamide
Formula:
C₆H₁₃NO MW: 115,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009047 CAS Number: 685-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...