N-Propylaniline 95%

Supplier: Apollo Scientific

OR920980-25G OR920980-5G
APOSOR920980-25GEA 285 EUR
APOSOR920980-25G APOSOR920980-5G
N-Propylaniline 95%
N-Propylaniline
Formula: C₉H₁₃N
MW: 135,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048678
CAS Number: 622-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR