(2,6-dimethylphenyl)methanol 95%

Supplier: Apollo Scientific

Synonyms: 2,6-Dimethylbenzyl alcohol

OR921010-10G OR921010-1G OR921010-5G
APOSOR921010-10GEA 108 EUR
APOSOR921010-10G APOSOR921010-1G APOSOR921010-5G
(2,6-dimethylphenyl)methanol 95%
(2,6-dimethylphenyl)methanol
Formula: C₉H₁₂O
MW: 136,194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128004
CAS Number: 62285-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR