2-Methylbutyraldehyde 95%

Supplier: Apollo Scientific
Danger

OR921296-100G OR921296-25G
APOSOR921296-100GEA 154 EUR
APOSOR921296-100G APOSOR921296-25G
2-Methylbutyraldehyde 95%
2-Methylbutyraldehyde
Formula: C₅H₁₀O
Boiling Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006984
CAS Number: 96-17-3
UN: 3371
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR