1-Butyl-1-phenylhydrazine 95%

Supplier: Apollo Scientific

OR921493-1G OR921493-25G OR921493-5G
APOSOR921493-1GEA 161 EUR
APOSOR921493-1G APOSOR921493-25G APOSOR921493-5G
1-Butyl-1-phenylhydrazine 95%
1-Butyl-1-phenylhydrazine
Formula: C₁₀H₁₆N₂
Storage Temperature: Ambient
MDL Number: MFCD03844777
CAS Number: 61715-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR