2-(1-Methyl-3-piperidinyl)-1-ethanol 95%
Supplier: Apollo Scientific
OR921640-1G
OR921640-250MG
APOSOR921640-1GEA
539
EUR
APOSOR921640-1G
APOSOR921640-250MG
2-(1-Methyl-3-piperidinyl)-1-ethanol 95%
2-(1-Methyl-3-piperidinyl)-1-ethanol
Formula:
C₈H₁₇NO MW: 143,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13183972 CAS Number: 101257-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...