Mesitylglyoxylic acid 95%

Supplier: Apollo Scientific

Synonyms: 2-Mesityl-2-oxoacetic acid

OR921873-5G
APOSOR921873-5GEA 137 EUR
APOSOR921873-5G
Mesitylglyoxylic acid 95%
Mesitylglyoxylic acid
Formula: C₁₁H₁₂O₃
MW: 192,2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151834
CAS Number: 3112-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR