2-Chloro-3-methoxybenzaldehyde 95%
Supplier: Apollo Scientific
OR921952-25G
OR921952-5G
APOSOR921952-25GEA
207
EUR
APOSOR921952-25G
APOSOR921952-5G
2-Chloro-3-methoxybenzaldehyde 95%
2-Chloro-3-methoxybenzaldehyde
Formula:
C₈H₇ClO₂ MW: 170,5930 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07369787 CAS Number: 54881-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...