3-Cyclopropyl-5-nitro-1H-pyrazole
Supplier: Apollo Scientific
|
Warning
|
OR922033-1G
OR922033-25G
OR922033-5G
APOSOR922033-1GEA
232
EUR
APOSOR922033-1G
APOSOR922033-25G
APOSOR922033-5G
3-Cyclopropyl-5-nitro-1H-pyrazole
3-Cyclopropyl-5-nitro-1H-pyrazole
Formula:
C₆H₇N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09701702 CAS Number: 326827-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...