Ethyl-3-cyclopropyl-2-oxopropanoate 95%
Supplier: Apollo Scientific
OR922077-1G
OR922077-250MG
APOSOR922077-1GEA
807
EUR
APOSOR922077-1G
APOSOR922077-250MG
Ethyl-3-cyclopropyl-2-oxopropanoate 95%
Ethyl-3-cyclopropyl-2-oxopropanoate
Formula:
C₈H₁₂O₃ MW: 156,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12407170 CAS Number: 64025-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...