3-Nitro-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Apollo Scientific

Synonyms: 3-Nitro-7-azaindole

OR922183-10G OR922183-1G OR922183-5G
APOSOR922183-10GEA 807 EUR
APOSOR922183-10G APOSOR922183-1G APOSOR922183-5G
3-Nitro-1H-pyrrolo[2,3-b]pyridine 95%
3-Nitro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅N₃O₂
MW: 163,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272225
CAS Number: 23709-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR