Phenyltrimethoxysilane 98%

Supplier: Apollo Scientific

Synonyms: Trimethoxy(phenyl)silane

OR922360-500G
APOSOR922360-500GEA 77 EUR
APOSOR922360-500G
Phenyltrimethoxysilane 98%
Phenyltrimethoxysilane
Formula: C₉H₁₄O₃Si
MW: 198,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025689
CAS Number: 2996-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR