3-Methyl-3-pentanol 98%

Supplier: Apollo Scientific

OR922588-100G OR922588-5G OR922588-25G
APOSOR922588-100GEA 417 EUR
APOSOR922588-100G APOSOR922588-5G APOSOR922588-25G
3-Methyl-3-pentanol 98%
3-Methyl-3-pentanol
Formula: C₆H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00004483
CAS Number: 77-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR