Methyl-p-tolylacetate 95%

Supplier: Apollo Scientific

Synonyms: Methyl 2-(p-tolyl)acetate, 4-Methylphenylacetic Acid Methyl Ester, Methyl 4-Methylphenylacetate, p-Tolylacetic Acid Methyl Ester

OR922797-25G
APOSOR922797-25GEA 59.6 EUR
APOSOR922797-25G
Methyl-p-tolylacetate 95%
Methyl-p-tolylacetate
Formula: C₁₀H₁₂O₂
MW: 164,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056243
CAS Number: 23786-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR