4-Ethylaniline 98%
Supplier: Apollo Scientific
OR922870-500G
APOSOR922870-500GEA
130
EUR
APOSOR922870-500G
4-Ethylaniline 98%
4-Ethylaniline
Formula:
C₈H₁₁N MW: 121,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007921 CAS Number: 589-16-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...