N-Methylsaccharin 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methylbenzo[d]isothiazol-3(2H)-one 1,1-dioxide

OR922935-1G OR922935-25G OR922935-5G
APOSOR922935-1GEA 147 EUR
APOSOR922935-1G APOSOR922935-25G APOSOR922935-5G
N-Methylsaccharin 98%
N-Methylsaccharin
Formula: C₈H₇NO₃S
MW: 197,21 g/mol
Melting Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00059263
CAS Number: 15448-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR