2-Phenoxypropanol 95%

Supplier: Apollo Scientific

OR923004-1G OR923004-25G OR923004-250MG OR923004-2.5G
APOSOR923004-1GEA 101 EUR
APOSOR923004-1G APOSOR923004-25G APOSOR923004-250MG APOSOR923004-2.5G
2-Phenoxypropanol 95%
2-Phenoxypropanol
Formula: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00142958
CAS Number: 4169-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR