N-(1,1,3,3-Tetramethylbutyl)formamide 95%

Supplier: Apollo Scientific

Synonyms: N-Formyl-1,1,3,3-tetramethylbutylamine, N-Formyl-tert-octylamine, tert-Octylformamide

OR923128-10G OR923128-1G OR923128-5G
APOSOR923128-10GEA 458 EUR
APOSOR923128-10G APOSOR923128-1G APOSOR923128-5G
N-(1,1,3,3-Tetramethylbutyl)formamide 95%
N-(1,1,3,3-Tetramethylbutyl)formamide
Formula: C₉H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD00039569
CAS Number: 10151-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR