o-Toluic acid 98%
Supplier: Apollo Scientific
Synonyms:
2-Methylbenzoic acid
OR923132-100G
OR923132-1KG
OR923132-500G
APOSOR923132-100GEA
125
EUR
APOSOR923132-100G
APOSOR923132-1KG
APOSOR923132-500G
o-Toluic acid 98%
o-Toluic acid
Formula:
C₈H₈O₂ MW: 136,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002477 CAS Number: 118-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...