4-(1-Ethylpropyl)pyridine 95%

Supplier: Apollo Scientific

Synonyms: 4-(1-Ethylpropyl)pyridine, 4-(3-Pentyl)pyridine, 3-(4-Pyridyl)pentane

OR923607-1G OR923607-25G OR923607-5G
APOSOR923607-1GEA 139 EUR
APOSOR923607-1G APOSOR923607-25G APOSOR923607-5G
4-(1-Ethylpropyl)pyridine 95%
4-(1-Ethylpropyl)pyridine
Formula: C₁₀H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD00059788
CAS Number: 35182-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR