Butyl-2-chloroethyl ether 95%

Supplier: Apollo Scientific

Synonyms: 1-(2-Chloroethoxy)butane, 2-Chloroethyl Butyl Ether

OR923720-25G OR923720-5G
APOSOR923720-25GEA 430 EUR
APOSOR923720-25G APOSOR923720-5G
Butyl-2-chloroethyl ether 95%
Butyl-2-chloroethyl ether
Formula: C₆H₁₃ClO
MW: 136,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059051
CAS Number: 10503-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR