4-Cyano-1-cyclohexene 95%

Supplier: Apollo Scientific

Synonyms: 1,2,3,6-Tetrahydrobenzonitrile, 3-Cyclohexene-1-carbonitrile

OR923732-5G
APOSOR923732-5GEA 145 EUR
APOSOR923732-5G
4-Cyano-1-cyclohexene 95%
4-Cyano-1-cyclohexene
Formula: C₇H₉N
MW: 107,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013778
CAS Number: 100-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR