2-(Methylthio)thiazole 98%

Supplier: Apollo Scientific

OR923771-100G
APOSOR923771-100GEA 166 EUR
APOSOR923771-100G
2-(Methylthio)thiazole 98%
2-(Methylthio)thiazole
Formula: C₄H₅NS₂
MW: 131,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00858981
CAS Number: 5053-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR