Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate 97%

Supplier: Apollo Scientific

Synonyms: 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylic acid Benzyl ester, N-Cbz-7-oxa-3-azabicyclo[4.1.0]heptane

OR923810-1G OR923810-500MG
APOSOR923810-1GEA 438 EUR
APOSOR923810-1G APOSOR923810-500MG
Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate 97%
Benzyl 7-Oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate
Formula: C₁₃H₁₅NO₃
MW: 233,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12407051
CAS Number: 66207-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR