3-Ethoxypropionic acid 95%
Supplier: Apollo Scientific
OR923879-5G
APOSOR923879-5GEA
52.2
EUR
APOSOR923879-5G
3-Ethoxypropionic acid 95%
3-Ethoxypropionic acid
Formula:
C₅H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00058997 CAS Number: 4324-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...