trans-4-Nitrocinnamaldehyde 95%

Supplier: Apollo Scientific

Synonyms: trans-3-(4-Nitrophenyl)-2-propenal, (E)-3-(4-Nitrophenyl)acrylaldehyde, (E)-4-Nitrocinnamaldehyde, (E)-3-(4-Nitrophenyl)-2-propenal

OR923923-1G
APOSOR923923-1GEA 56.7 EUR
APOSOR923923-1G
trans-4-Nitrocinnamaldehyde 95%
trans-4-Nitrocinnamaldehyde
Formula: C₉H₇NO₃
MW: 177,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007379
CAS Number: 49678-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR