1-Methyl-1H-pyrazole-5-carbonitrile 95%

Supplier: Apollo Scientific

OR924140-1G
APOSOR924140-1GEA 69.6 EUR
APOSOR924140-1G
1-Methyl-1H-pyrazole-5-carbonitrile 95%
1-Methyl-1H-pyrazole-5-carbonitrile
Formula: C₅H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD09701708
CAS Number: 66121-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR